Learn More
BRD4 Antibody (CL1115) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52958AF532
Product Code. 30305383
  • £558.00 / 0.10mL
Description
BRD4 Monoclonal antibody specifically detects BRD4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BRD4
Monoclonal
Alexa Fluor 532
bromodomain containing 4, bromodomain-containing 4, CAPchromosome-associated protein, HUNK1bromodomain-containing protein 4, HUNKI, MCAP, Protein HUNK1
This antibody was developed using a recombinant protein derived from O60885, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Signal Transduction
23476
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL1115
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified