missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11859-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene was identified based on its homology to the gene encoding the RING3 protein, a serine/threonine kinase. The gene localizes to 9q34, a region which contains several major histocompatibility complex (MHC) genes. The function of the encoded protein is not known.Specifications
| BRD3 | |
| Polyclonal | |
| Unconjugated | |
| BRD3 | |
| BRD3, bromodomain containing 3, KIAA0043, ORFX, RING3 like protein, RING3L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362092, 67382, 8019 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15059, Q8K2F0 | |
| Brd3 | |
| BRD3 Fusion Protein Ag2433 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction