missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BRD3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16311045
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16311045 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16311045 Supplier Invitrogen™ Supplier No. PA584088

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MPQEEVELLP PAPKGKGRKP AAGAQSAGTQ QVAAVSSVSP ATPFQSVPPT VSQTPVIAAT PVPTITANVT SV Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 90%.

This gene was identified based on its homology to the gene encoding the RING3 protein, a serine/threonine kinase. The gene localizes to 9q34, a region which contains several major histocompatibility complex (MHC) genes. The function of the encoded protein is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD3
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BRD3
Gene Accession No. Q15059
Gene Alias 2410084F24Rik; AW060456; BRD3; bromodomain containing 3; bromodomain containing 3; bromodomain-containing protein 3; bromodomain containing protein 3; bromodomain-containing 3; bromodomain-containing FSH-like protein FSRG2; Bromodomain-containing protein 3; bromodomain-containing protein 3 short isoform; Fsrg2; KIAA0043; ORFX; RING3L; RING3-like protein; RINGL3
Gene Symbols BRD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BRD3. Recombinant protein control fragment (Product #RP-101454).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8019
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.