missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £318.00
Specifications
| Antigen | BRD2 |
|---|---|
| Dilution | Western Blot 1:1000, ELISA 1:100 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18312775
|
Novus Biologicals
NBP3-18662-25UL |
25 μg |
£139.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18375322
|
Bio-Techne
NBP3-18662-100UL |
100 μg |
£318.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BRD2 Polyclonal antibody specifically detects BRD2 in Mouse samples. It is validated for Western Blot, ELISASpecifications
| BRD2 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, DNA Repair, Protein Kinase | |
| Whole antiserum | |
| bromodomain containing 2, bromodomain-containing 2, bromodomain-containing protein 2, D6S113E, female sterile homeotic-related gene 1, FSRG1FSH, KIAA9001FLJ31942, NAT, O27.1.1, Really interesting new gene 3 protein, RING3DKFZp686N0336, RNF3 | |
| Polyclonal antibody raised in rabbit against mouse BRD2 (bromodomain containing 2), using a KLHconjugated synthetic peptide containing an amino acid sequence from the central part of the protein (1). | |
| Whole Antiserum |
| Western Blot 1:1000, ELISA 1:100 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Mouse | |
| P25440 | |
| 6046 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title