missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BRD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109455
This item is not returnable.
View return policy
Description
Immunogen sequence: MLQNVTPHNK LPGEGNAGLL GLGPEAAAPG KRIRKPSLLY EGFESPTMAS VPALQLTPAN PPPPEVSNPK KPGRVTNQLQ YLHKVVMKAL WKHQFAWPFR.
This gene encodes a transcriptional regulator that belongs to the BET (bromodomains and extra terminal domain) family of proteins. This protein associates with transcription complexes and with acetylated chromatin during mitosis, and it selectively binds to the acetylated lysine-12 residue of histone H4 via its two bromodomains. The gene maps to the major histocompatibility complex (MHC) class II region on chromosome 6p21. 3, but sequence comparison suggests that the protein is not involved in the immune response. This gene has been implicated in juvenile myoclonic epilepsy, a common form of epilepsy that becomes apparent in adolescence. Multiple alternatively spliced variants have been described for this gene, but the full-length nature of some of these variants has not been determined.
Specifications
| BRD2 | |
| Polyclonal | |
| Unconjugated | |
| BRD2 | |
| AW228947; Brd2; bromodomain containing 2; bromodomain-containing 2; bromodomain-containing protein 2; D17H6S113E; D6S113E; DADB-17J1.3; DKFZp686N0336; female sterile homeotic-related gene 1; Female sterile homeotic-related protein 1; FLJ31942; Frg-1; FSH; Fsrg1; Fsrg-1; Kiaa4005; KIAA9001; mKIAA4005; Nat; O27.1.1; Protein RING3; Really interesting new gene 3 protein; RING3; RING3 protein; Rnf3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6046 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.33 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P25440 | |
| BRD2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human BRD2 (NP_0050951). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction