missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD2 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33404-100ul
This item is not returnable.
View return policy
Description
BRD2 Monoclonal antibody specifically detects BRD2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| BRD2 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| bromodomain containing 2, bromodomain-containing 2, bromodomain-containing protein 2, D6S113E, female sterile homeotic-related gene 1, FSRG1FSH, KIAA9001FLJ31942, NAT, O27.1.1, Really interesting new gene 3 protein, RING3DKFZp686N0336, RNF3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-345 of human BRD2 (NP_005095.1).,, Sequence:, MLQNVTPHNKLPGEGNAGLLGLGPEAAAPGKRIRKPSLLYEGFESPTMASVPALQLTPANPPPPEVSNPKKPGRVTNQLQYLHKVVMKALWKHQFAWPFRQPVDAVKLGLPDYHKIIKQPMDMGTIKRRLENNYYWAASECMQDFNTMFTNCYIYNKPTDDIVLMAQTLEKIFLQKVASMPQEEQELVVTIPKNSHKKGAKLAALQGSVTSAHQVPAVSSVSHTALYTPPPEIPTTVLNIPHPSVISSPLLKSLHSAGPPLLAVTAAPPAQPLAKKKGVKRKADTTTPTPTAILAPGSPASPPGSLEPKAARLPPMRRESGRPIKPPRKDLPDSQQQHQSSKKGK | |
| 100 μL | |
| Cellular Markers, DNA Repair, Protein Kinase | |
| 6046 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction