missing translation for 'onlineSavingsMsg'
Learn More

BRD2 Antibody (JG40-42), Novus Biologicals™

Product Code. 18358610 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358610 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358610 Supplier Novus Biologicals Supplier No. NBP275422

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BRD2 Monoclonal specifically detects BRD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JG40-42
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:100-1:400
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias bromodomain containing 2, bromodomain-containing 2, bromodomain-containing protein 2, D6S113E, female sterile homeotic-related gene 1, FSRG1FSH, KIAA9001FLJ31942, NAT, O27.1.1, Really interesting new gene 3 protein, RING3DKFZp686N0336, RNF3
Gene Symbols BRD2
Host Species Rabbit
Immunogen Synthetic peptide within Human BRD2 aa 1-50/ 801. (SwissProt: P25440 Human)
Molecular Weight of Antigen 88 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cellular Markers, DNA Repair, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6046.0
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.