missing translation for 'onlineSavingsMsg'
Learn More

BRD2 Antibody (BL-167-2A2), Novus Biologicals™

Product Code. 18377190 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377190 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377190 Supplier Novus Biologicals Supplier No. NBP276410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BRD2 Monoclonal specifically detects BRD2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD2
Applications Western Blot, Immunoprecipitation, Immunoassay
Classification Monoclonal
Clone BL-167-2A2
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunoprecipitation 20 ul/1mg lysate
Formulation Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA with 0.09% Sodium Azide
Gene Alias bromodomain containing 2, bromodomain-containing 2, bromodomain-containing protein 2, D6S113E, female sterile homeotic-related gene 1, FSRG1FSH, KIAA9001FLJ31942, NAT, O27.1.1, Really interesting new gene 3 protein, RING3DKFZp686N0336, RNF3
Gene Symbols BRD2
Host Species Rabbit
Immunogen Immunogen sequence between amino acids 675 and 725
Purification Method Antigen Affinity-purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 6046.0
Target Species Human, Mouse
Content And Storage Store at 2-8°C/ 1 year from date of receipt
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.