Learn More
Invitrogen™ BRD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597274
Description
Immunogen sequence: MRRKGRCHRG SAARHPSSPC SVKHSPTRET LTYAQAQRMV EIEIEGRLHR ISIFDPLEII LEDDLTAQEM SECNSNKENS ERPPVCLRTK RHKNNRVKKK NEALPSAHGT PASASALPEP KVRIVEYSPP SAPRRPPVYY KFIEKSAEEL DNEVEYDMDE EDYAWLEIVN EKRKGDCVPA VSQSMFEFLM DRFEKESHCE NQKQGEQQSL IDEDAVCCIC MDGECQNSNV ILFCDMCNLA VHQECYGVPY; Positive Samples: HeLa, Jurkat; Cellular Location: Nucleus.
This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types.
Specifications
| BRD1 | |
| Polyclonal | |
| Unconjugated | |
| BRD1 | |
| 1110059H06Rik; AI316859; BR140-like protein; BRD1; BRL; Bromodomain and PHD finger-containing protein 2; bromodomain containing 1; bromodomain-containing protein 1; BRPF1; BRPF2; DKFZp686F0325; mKIAA4191; RP3-522J7.4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23774 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.54 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95696 | |
| BRD1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human BRD1 (NP_0553921). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.