missing translation for 'onlineSavingsMsg'
Learn More

BRD 4354, MedChemExpress

Product Code. 16497397
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497397 5 mg 5mg
16495516 100 mg 100mg
16453196 25 mg 25mg
16444688 10 mg 10mg
16440977 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497397 Supplier Medchem Express Supplier No. HY112719
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BRD 4354 is a moderately potent inhibitor of HDAC5 and HDAC9, with IC50s of 0.85 and 1.88 μM, respectively.

BRD 4354 is a moderately potent inhibitor of HDAC5 and HDAC9, with IC50s of 0.85 and 1.88 μM, respectively.

Specifications

Chemical Name or Material BRD 4354
CAS 315698-07-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H23ClN4O
Quantity 5 mg
Solubility Information DMSO : 12.5 mg/mL (32.65 mM; Need ultrasonic)
SMILES OC1=C2N=CC=CC2=C(Cl)C=C1C(N3CCN(CC)CC3)C4=CC=CN=C4
Molecular Weight (g/mol) 382.89
Formula Weight 382.89
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.