missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BRD 0539
GREENER_CHOICE

Product Code. 18776956
Click to view available options
Quantity:
2 mg
10 mg
Unit Size:
10mg
2mg
This item is not returnable. View return policy

Product Code. 18776956

Brand: Tocris Bioscience™ 7153/2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell permeable and reversible Cas9 inhibitor

BRD 0539 is a cell-permeable and reversible SpCas9 (Streptococcus pyogenes Cas9) inhibitor. BRD 0539 shows dose-dependent SpCas9 inhibition in an in vitro DNA cleavage assay (apparent IC50 = 22 μM) and in an eGFP disruption assay (apparent EC50 = 11 μM). Allows dose and temporal control of SpCas9-based systems.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3 aR,4 R,9 bR)-8-(2-Fluorophenyl)-2,3,3 A,4,5,9 b-hexahydro-1-[(4-methylphenyl)sulfonyl]-1 H-pyrrolo[3,2-c]quinoline-4-methanol
CAS 1403838-79-8
Quantity 2 mg
Target CRISPR Reagents
Molecular Formula C25H25FN2O3S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.