missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BRD 0539

Click to view available options
Quantity:
2 mg
10 mg
Unit Size:
10mg
2mg
Description
BRD 0539 is a cell-permeable and reversible SpCas9 (Streptococcus pyogenes Cas9) inhibitor. BRD 0539 shows dose-dependent SpCas9 inhibition in an in vitro DNA cleavage assay (apparent IC50 = 22 μM) and in an eGFP disruption assay (apparent EC50 = 11 μM). Allows dose and temporal control of SpCas9-based systems.
Specifications
Specifications
| Chemical Name or Material | (3 aR,4 R,9 bR)-8-(2-Fluorophenyl)-2,3,3 A,4,5,9 b-hexahydro-1-[(4-methylphenyl)sulfonyl]-1 H-pyrrolo[3,2-c]quinoline-4-methanol |
| CAS | 1403838-79-8 |
| Quantity | 10 mg |
| Target | CRISPR Reagents |
| Molecular Formula | C25H25FN2O3S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction