missing translation for 'onlineSavingsMsg'
Learn More

BRCAA1 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18409696 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409696 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409696 Supplier Novus Biologicals Supplier No. NBP126619C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BRCAA1 Polyclonal antibody specifically detects BRCAA1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCAA1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 180 kDa Sin3-associated polypeptide, ARID domain-containing protein 4B, AT rich interactive domain 4B (RBP1- like), AT rich interactive domain 4B (RBP1-like), AT-rich interactive domain-containing protein 4B, BRCAA1DKFZp313M2420, breast cancer-associated antigen 1, Breast cancer-associated antigen BRCAA1, breast carcinoma-associated antigen, Histone deacetylase complex subunit SAP180, Rb-binding protein homolog, RBBP1L1RBP1L1BCAA, RBP1-like protein, retinoblastoma binding protein 1-like 1, Retinoblastoma-binding protein 1-like 1, SAP180MGC163290, SIN3A-associated protein 180, Sin3-associated polypeptide p180
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 450 and 500 of human AT rich interactive domain 4B using the numbering given in entry NP_057458.4 (GeneID 51742).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 51742
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.