missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19791-1-AP-150UL
This item is not returnable.
View return policy
Description
Inherited mutations in BRCA1 and this gene, BRCA2, confer increased lifetime risk of developing breast or ovarian cancer. Both BRCA1 and BRCA2 are involved in maintece of genome stability, specifically the homologous recombination pathway for double-strand DNA repair. The BRCA2 protein contains several copies of a 70 aa motif called the BRC motif, and these motifs mediate binding to the RAD51 recombinase which functions in DNA repair. BRCA2 is considered a tumor suppressor gene, as tumors with BRCA2 mutations generally exhibit loss of heterozygosity (LOH) of the wild-type allele.Specifications
| BRCA2 | |
| Polyclonal | |
| Unconjugated | |
| BRCA2 | |
| BRCA2, BRCC2, breast cancer 2, early onset, BROVCA2, FACD, FAD, FAD1, FANCB, FANCD, FANCD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12190, 360254, 675 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35923, P51587, P97929 | |
| Brca2 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction