missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BRCA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-28966
This item is not returnable.
View return policy
Description
BRCA2 Polyclonal antibody specifically detects BRCA2 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| BRCA2 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| BRCA1/BRCA2-containing complex, subunit 2, BRCC2, breast and ovarian cancer susceptibility gene, early onset, breast cancer 2 tumor suppressor, breast cancer 2, early onset, breast cancer susceptibility protein BRCA2, breast cancer type 2 susceptibility protein, BROVCA2, FACD, FAD, FAD1, FANCB, FANCD, FANCD1Fanconi anemia, complementation group D1, Fanconi anemia group D1 protein, GLM3, PNCA2 | |
| between 450 and 500 | |
| 100 μg | |
| Breast Cancer, Cancer | |
| 675 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction