missing translation for 'onlineSavingsMsg'
Learn More

BRCA2 Antibody (234403) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30735180 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735180 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735180 Supplier Novus Biologicals Supplier No. FAB2476MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BRCA2 Monoclonal antibody specifically detects BRCA2 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCA2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 234403
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias BRCA1/BRCA2-containing complex, subunit 2, BRCC2, breast and ovarian cancer susceptibility gene, early onset, breast cancer 2 tumor suppressor, breast cancer 2, early onset, breast cancer susceptibility protein BRCA2, breast cancer type 2 susceptibility protein, BROVCA2, FACD, FAD, FAD1, FANCB, FANCD, FANCD1Fanconi anemia, complementation group D1, Fanconi anemia group D1 protein, GLM3, PNCA2
Host Species Mouse
Immunogen E. coli-derived recombinant human BRCA2, Met1-Thr200, Accession # P51587
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 675
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.