missing translation for 'onlineSavingsMsg'
Learn More

BRCA1 Antibody Pack, Novus Biologicals™

Product Code. 18534831 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Pack
Unit Size:
1 set
Product Code. Quantity unitSize
18534831 1 Pack 1 set
1 options
This item is not returnable. View return policy

Product Code. 18534831

Brand: Novus Biologicals NB1009361Pack

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pack

BRCA1 Antibody Packantibody specifically detects BRCA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCA1
Applications Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Conjugate Unconjugated
Gene Alias BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1
Immunogen See individual datasheets.
Quantity 1 Pack
Regulatory Status RUO
Research Discipline Breast Cancer
Gene ID (Entrez) 672
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.