missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCA1 Antibody (MU), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-600H
This item is not returnable.
View return policy
Description
BRCA1 Monoclonal specifically detects BRCA1 in Human samples. It is validated for Western Blot.
Specifications
| BRCA1 | |
| Monoclonal | |
| HRP | |
| P38398 | |
| BRCA1 | |
| Human BRCA1 corresponding to residues 1314-1864 [UniProt# P38398] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MU | |
| Western Blot | |
| BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1 | |
| Mouse | |
| 240 kDa | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
| 672 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction