missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCA1 Antibody (BRCA1/1472), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54337UV
This item is not returnable.
View return policy
Description
BRCA1 Monoclonal antibody specifically detects BRCA1 in Human samples. It is validated for ELISA, Protein Array, CyTOF
Specifications
| BRCA1 | |
| Monoclonal | |
| DyLight 350 | |
| BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1 | |
| Recombinant fragment (around aa 445-620) of human BRCA1 protein (exact sequence is proprietary) (Uniprot: P38398) | |
| 100 μg | |
| Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
| 672 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Protein Array, CyTOF | |
| BRCA1/1472 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction