Learn More
Endecotts™ Brass Lid
Product Code. 15339232
Click to view available options
Diameter (Metric):
300 mm
Product Code. 15339232
Brand: Endecotts™ 300BRASLID
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with Endecotts™ sieves. Endecotts™ Brass Lids are widely used with test sieves for all types of laboratory sampling and particle size analysis.
  • Brass construction
  • Can be used with either full or half height sieves
  • Available in a wide variety of diameters
Specifications
Product Type Lid
Material Brass
Diameter (Metric) 300 mm
Packaging Quantity 1 Ea.