Learn More
Brass gauze, alloy 260, 80 mesh woven from 0.14mm (0.0055 in.) dia. wire; Cu:Zn; 70:30 wt%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 045187.HB
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Brass gauze, Alloy 260 | |
Cu:Cn 70:30 wt% | |
300 x 300 mm | |
Odorless |
Woven from 0.14mm (0.0055 in.) dia. wire | |
MFCD00171255 | |
80 Mesh |