Learn More
Gas Arc™ Inline Flashback Arrestor
Product Code. 12950820
Encompass
Click to view available options
For Use With (Application):
Fuel Gas
Oxygen
Unit Size:
Each
Product Code. 12950820
Brand: Gas Arc™ FBA07051
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Connection Type 3/8 BSP RH Female (inlet); 3/8 BSP RH Male (outlet)
Type Inline Flashback Arrestor
For Use With (Application) Oxygen
Pressure 20 bar (inlet)/20 bar (outlet)