Learn More
Brass foil, alloy 260, 0.51mm (0.02 in.) thick, 15x45cm (5.9x18in), Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 013502.KJ
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Brass foil, Alloy 260 | |
MFCD00171255 | |
Odorless |
0.51mm (0.02 in.) thick, 15x45cm (5.9x18 in.) | |
5 Pc. | |
Foil |