missing translation for 'onlineSavingsMsg'
Learn More

Brass foil, alloy 260, 0.3mm (0.012in) thick

Product Code. 15482167
Click to view available options
Quantity:
150 x 150 mm
300 x 300 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15482167

Brand: Thermo Scientific Alfa Aesar 045135.HB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Brass foil, alloy has a wide range of applications, such as appliance parts, highly formable parts, ammunition components, decorative parts of furniture and cabinets, name plates, engraving plates, overlays, legend plates, lamp fixtures, fasteners, locks, hinges, flashlight shells, connectors, fittings, circuit board relays, electronic components, switches, nuts, bolts, valve stems and valve components, radiator cores and tanks, brackets, frames, fixtures, ornamental works, and used extensively in the musical instrument, electrical equipment, plumbing, central heating, seawater lines and desalination fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brass foil, Alloy 260
Name Note 0.3mm (0.012 in.) thick
Quantity 300 x 300 mm
Odor Odorless
Form Foil
MDL Number MFCD00171255
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.