missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BRAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566210
This item is not returnable.
View return policy
Description
Immunogen sequence: STPNQYMVLIK FRAQADADSF YMTCNGRQFN SIEDDVCQLV YVERAEVLKS EDGASLPVMD LTELPKCTVC LERMDESVNG ILTTLCNHSF HSQC Highest antigen sequence identity to the following orthologs - mouse 98%, rat 97%.
Negatively regulates MAP kinase activation by limiting the formation of Raf/MEK complexes probably by inactivation of the KSR1 scaffold protein. Also acts as a Ras responsive E3 ubiquitin ligase that, on activation of Ras, is modified by auto-polyubiquitination resulting in the release of inhibition of Raf/MEK complex formation. May also act as a cytoplasmic retention protein with a role in regulating nuclear transport.
Specifications
| BRAP | |
| Polyclonal | |
| Unconjugated | |
| BRAP | |
| 3010002G07Rik; BRAP; BRAP2; BRCA1 associated protein; BRCA1-associated protein; galectin-2-binding protein; IMP; Impedes mitogenic signal propagation; renal carcinoma antigen NY-REN-63; RING finger protein 52; RING-type E3 ubiquitin transferase BRAP2; RNF52 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8315 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z569 | |
| BRAP | |
| Recombinant Human BRAP. Recombinant protein control fragment (Product #RP-106892). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction