missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-42696
This item is not returnable.
View return policy
Description
BRAP Polyclonal specifically detects BRAP in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| BRAP | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| BRAP | |
| The immunogen for this product maps to a region between residue 560 and 592 of human BRCA1 associated protein using the numbering given in entry NP_006759.3 (GeneID 8315). | |
| 100 μL | |
| Lipid and Metabolism | |
| 8315 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| BRAP2RING finger protein 52, BRCA1 associated protein, EC 6.3.2.-, IMPBRCA1-associated protein, Impedes mitogenic signal propagation, Renal carcinoma antigen NY-REN-63, RNF52galectin-2-binding protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction