missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VACUUBRAND™ VHCpro System Accessory - 8-channel Pipet Tip Adapter
Shop All Vacuubrand Products
Click to view available options
For Use With (Equipment):
VHCPro
Unit Size:
Each
Description
- Includes tip ejector
Specifications
Specifications
| Product Type | 8 Channel Aspirator |
| Autoclavable | Autoclavable |
| Color | Black/White |
| Channels | 8 |
| Material | Polypropylene |
| For Use With (Application) | Aspiration from multi-well plates |
| For Use With (Equipment) | VHCPro |
| Includes | 8-channel manifold only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction