missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Transferpette™ S Single Channel Fixed Volume Pipet

Product Code. 10308832 Shop All Brand Products
Change view
Click to view available options
Accuracy:
±0.6%
±0.8%
±1%
Model:
F10
F100
F1000
F20
F200
F25
F50
F500
Volume (Metric):
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
500 μL
1000 μL
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Accuracy Model Volume (Metric) unitSize
10308832 ±0.6% F500 500 μL Each
10454442 ±0.8% F50 50 μL Each
10165432 ±1% F10 10 μL Each
10319022 ±0.8% F20 20 μL Each
10759463 ±0.8% F25 25 μL Each
10695453 ±0.6% F100 100 μL Each
10464442 ±0.6% F200 200 μL Each
10635463 ±0.6% F1000 1000 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10308832 Supplier BRAND™ Supplier No. 704754

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Uses innovative and ergonomic one-handed operation.Transferpette™ S Single Channel Fixed Volume Pipet offers robust yet lightweight construction. Easy Calibration™ technology provides long lasting reliability, accuracy, and precision

Device is lightweight with one-handed volume adjustment.

  • Centralized pipetting button with optimized stroke length
  • Easy in-lab service
  • Robust and lightweight
  • Accepts universal tips
  • Autoclavable at 121°C/250°F (20 minutes)

TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±0.6%
Certifications/Compliance ISO9001, CE, IVD, certificate of performance, individual calibration certificate
Compatible Tips 50 to 1000μL tip, 50 to 1250μL tip
Autoclavable Autoclavable
For Use With 50 μL to 1000 μL Tip, 50 μL to 1250 μL Tip
Model F500
Number of Channels 1
Type Fixed Volume Pipette
Volume (Metric) 500 μL
For Use With (Equipment) 50 to 1000μL tip, 50 to 1250μL tip
Includes Centralized piston button design, shelf mount, 4 digit display, steam sterilizable shaft, operating manual
Warranty 3 Years
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.