Learn More
BRAND™ Transferpette™ S Adjustable Single Channel Pipet Starter Sets
Product Code. 10161623 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
0.1 to 1, 0.5 to 10, 10 to 100 μL
0.5 to 10, 10 to 100, 100 to 1000 μL
100 to 1000, 500 to 5000, 1000 to 10,000 μL
2 to 20, 20 to 200, 100 to 1000 μL
Includes:
D-1, D-10, D-100 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
D-10, D-100, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
D-20, D-200, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
D1000, D5000, D10000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
Type:
Macro Set
Micro Set
Pipette Set, Standard
Starter Set, Adjustable
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Includes Type unitSize
10161623 0.1 to 1, 0.5 to 10, 10 to 100 μL D-1, D-10, D-100 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes Micro Set Each
10179002 2 to 20, 20 to 200, 100 to 1000 μL D-20, D-200, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes Starter Set, Adjustable Each
10715883 100 to 1000, 500 to 5000, 1000 to 10,000 μL D1000, D5000, D10000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes Macro Set Each
10563815 0.5 to 10, 10 to 100, 100 to 1000 μL D-10, D-100, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes Pipette Set, Standard Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10161623 Supplier BRAND™ Supplier No. 704790
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Set contents:
  • Three adjustable volume pipettes
  • Three shelf/rack mounts
  • Three filled polypropylene Tip Boxes
Simple one-handed operation units exhibits high degree accuracy, precision, maximum versatility and reliable performance with Easy Calibration™ technology
  • Available as Standard Set, Micro Set, Midsize Set and Macro Set
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
For Use With Laboratory
Number of Channels 1
Product Line Transferpette
Type Micro Set
Volume (Metric) 0.1 to 1, 0.5 to 10, 10 to 100 μL
For Use With (Application) Demanding laboratory applications
Includes D-1, D-10, D-100 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes