Learn More
BRAND™ Transferpette™ S Multichannel Pipet
Protects against accidental volume changes using innovative one-handed volume adjustment with brake. Supplied with performance certificate, operating manual, shelf mount, two tip boxes, reagent reservoir, pack of replacement v-rings, and one year warranty.
Brand: BRAND™ 703726
Code : 03
Additional Details : Weight : 0.57000kg
Includes: Shelf/rack mount, box of pipette tips with two refills, set of replacement V-rings, certificate of performance and comprehensive operating manual
Description
Protects against accidental volume changes using innovative one-handed volume adjustment with brake. BrandTech™ Transferpette™ S Multichannel Pipet is supplied with performance certificate, operating manual, shelf mount, two tip boxes, reagent reservoir, pack of replacement v-rings, and one year warranty. Patented individually removable/replaceable tip cones provide easy in-lab cleaning.
Device has one-handed volume adjustment with brake for secure setting.
- With central button control layout
- Short, 12.5mm piston stroke-length
- V-rings for low-force tip mounting and ejection
- Four digit volume display
- Lightweight
- Individually removable/replaceable patented tip cones
- Autoclavable at 121°C (20 minutes)
Specifications
±0.8% | |
12 | |
Pipette | |
Digital | |
12.5mm | |
241.3 x 142.8 x 123.8mm | |
241.3 mm | |
142.8 mm | |
123.8 mm |
Autoclavable | |
Transferpette™ S | |
5 to 50 μL | |
Shelf/rack mount, box of pipette tips with two refills, set of replacement V-rings, certificate of performance and comprehensive operating manual | |
1 Year | |
9.5 in. | |
5.62 in. | |
4.87 in. |