missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Self-adhesive Plate Sealing Film

Product Code. 10135371 Shop All Brand Products
Change view
Click to view available options
Material:
Aluminum
Polyester
Polyethylene, Polypropylene
Polypropylene
Rayon
Vinyl
For Use With (Application):
Automation
Cell and tissue culture
Cold storage
ELISA, PCR
ELISA, Real-time PCR
Luminescence measurements
Storage
Storage, fluorescence measurements
No. per Pack:
100
100 Sheets
50 Sheets
Unit Size:
Pack of 100
Pack of 50
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material For Use With (Application) No. per Pack unitSize
10135371 Rayon Cell and tissue culture 100 Sheets Pack of 100
10655861 Polyester ELISA, PCR 100 Sheets Pack of 100
11673119 Polypropylene Storage 100 Sheets Pack of 100
10767564 Polyester ELISA, Real-time PCR 100 Sheets Pack of 100
10182962 Aluminum Cold storage 100 Sheets Pack of 100
10599892 Rayon Cell and tissue culture 50 Sheets Pack of 50
10135623 Polyethylene, Polypropylene Automation 50 Sheets Pack of 50
10185763 Vinyl Storage, fluorescence measurements 50 Sheets Pack of 50
10145813 Vinyl Luminescence measurements 50 Sheets Pack of 50
15660499 Vinyl Automation 100 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 10135371 Supplier BRAND™ Supplier No. 701364
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.

  • Easily apply to plates and remove without the use of expensive equipment.
  • Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
  • Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
    • Polyethylene
    • Polyester
    • Aluminum
    • Rayon
    • Polypropylene
    • Vinyl
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Description Gas permeable
Material Rayon
Sterility Non-sterile
Autoclavable Not Autoclavable
No. per Pack 100 Sheets
Evaporation Barrier No
For Use With (Application) Cell and tissue culture
Pre-slit No
Product Type Sealing Film
Adhesive Pressure Sensitive
Thickness 10.1 mil.
Pierceable Yes
Temperature Range (Metric) -20°C to +80°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.