missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Individual Stand for Transferpette™ Electronic Pipets

Product Code. 10327682 Shop All Brand Products
Change view
Click to view available options
For Use With (Equipment):
Transferpette Electronic Single Channel Pipette (500 to 5000 μL)
Transferpette Electronic Single Channel Pipette up to 1000μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10327682 Transferpette Electronic Single Channel Pipette (500 to 5000 μL) Each
10620853 Transferpette Electronic Single Channel Pipette up to 1000μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10327682 Supplier BRAND™ Supplier No. 705386
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Also suitable for corresponding models of Transferpette™ and Transferpette S pipets.

  • Available for two different capacities
  • Provided individually
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Transparent/White
Disposable Reusable
For Use With Transferpette/Transferpette S
Material Polypropylene, Stainless Steel
Number of Channels 1
Sterility Non-sterile
Type Pipette Stand
For Use With (Equipment) Transferpette Electronic Single Channel Pipette (500 to 5000 μL)
Width (English) 4.5 in.
Height (English) 3.12 in.
Height (Metric) 7.92 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.