missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ VITLAB™ Reagent Bottles with Stoppers

Product Code. 15792036
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Unit Size:
Each
Pack of 10
Pack of 20
This item is not returnable. View return policy

Product Code. 15792036

Brand: BRANDTECH™ VITLAB™ 100494

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

VITLAB polypropylene reagent bottles with polypropylene stoppers have good chemical resistance. Autoclavable at 121 C for 20 minutes.

  • NS stoppers and good chemical resistance for long-term storage of liquids.
  • Autoclavable at 121 C, according to DIN EN 285

Specifications

Capacity 8.45 oz., 250mL
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Transparent
Diameter (English) 2.75 in.
Material Polypropylene
Quantity 20/Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 138 mm
Product Type Reagent Bottle
Height (English) 5.43 in.
Closure Material Polypropylene
Closure Size 19/26 mm
Diameter (Metric) 70 mm
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.