missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ VITLAB™ Reagent Bottles with Stoppers

Product Code. 15782036
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Unit Size:
Each
Pack of 10
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15782036 100 mL Pack of 20
10396041 250 mL Each
15792036 250 mL Pack of 20
15702046 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15782036 Supplier BRANDTECH™ VITLAB™ Supplier No. 100394
Add to basket
Add to basket
This item is not returnable. View return policy

VITLAB polypropylene reagent bottles with polypropylene stoppers have good chemical resistance. Autoclavable at 121 C for 20 minutes.

  • NS stoppers and good chemical resistance for long-term storage of liquids.
  • Autoclavable at 121 C, according to DIN EN 285

Specifications

Capacity 3.38 oz., 100mL
Capacity (Metric) 100 mL
Capacity (English) 3.38 oz.
Color Transparent
Material Polypropylene
Quantity 20/Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Product Type Reagent Bottle
Closure Material Polypropylene
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.