missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Accessories for PLT™ (Pipette Leak Testing) Units

Product Code. p-4445487 Shop All Brand Products
Click to view available options
Type:
Multichannel Pipet Adapter
Replacement Filters
Single Channel Pipette Adapter
Universal Power Supply
For Use With (Application):
Testing with Tips
Testing with or without Tips
Testing without Tips
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 11683239

Brand: BRAND™ 703977

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with BRAND™ PLT™ unit. These accessories are designed for improved process reliability in identifying even the smallest leaks in your air displacement pipette.

Available accessories include:

  • Part No. 703978: Polyethylene filters and cap for pipette adapters, includes ten filters and one cap.
  • Part No. 703977: Four-channel pipette adapter, for testing with and without tips, includes four plugs.
  • Part No. 703975: Single-channel pipette adapter for testing with tips, includes one plug.
  • Part No. 703976: Single-channel pipette adapter for testing without tips, includes one plug.
  • Part No. 703979: Universal AC Adapter, 100-240V, 50/60Hz.
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips All multi-channel tips
Description Multichannel Pipet Adapter, Includes four plugs
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Black
Disposable Reusable
For Use With Multichannel Pipettes/PLT Unit
Material Silicone
Number of Channels 4
Sterility Non-sterile
Type Multichannel Pipet Adapter
For Use With (Application) Testing with or without Tips
For Use With (Equipment) PLT Pipette Leak Testing Unit
Includes 4 Plugs
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.