missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Accessories for PLT™ (Pipette Leak Testing) Units

Product Code. 11693239 Shop All Brand Products
Change view
Click to view available options
Type:
Multichannel Pipet Adapter
Replacement Filters
Single Channel Pipette Adapter
Universal Power Supply
For Use With (Application):
Testing with Tips
Testing with or without Tips
Testing without Tips
Unit Size:
Each
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Application) Unit Size
11693239 Single Channel Pipette Adapter Testing with Tips Each
11683239 Multichannel Pipet Adapter Testing with or without Tips Each
11648419 Replacement Filters - Pack of 10
11673239 Single Channel Pipette Adapter Testing without Tips Each
11633239 Universal Power Supply - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11693239 Supplier BRAND™ Supplier No. 703975
  • ONLINE EXCLUSIVE PRICE
    £38.70
    £35.63 / Pack of 1
    Save £3.07
    7% Off
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with BRAND™ PLT™ unit. These accessories are designed for improved process reliability in identifying even the smallest leaks in your air displacement pipette.

Available accessories include:

  • Part No. 703978: Polyethylene filters and cap for pipette adapters, includes ten filters and one cap.
  • Part No. 703977: Four-channel pipette adapter, for testing with and without tips, includes four plugs.
  • Part No. 703975: Single-channel pipette adapter for testing with tips, includes one plug.
  • Part No. 703976: Single-channel pipette adapter for testing without tips, includes one plug.
  • Part No. 703979: Universal AC Adapter, 100-240V, 50/60Hz.
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips All tips
Description Single Channel Pipette Adapter, For testing with tips, Includes plugs
Autoclavable Not Autoclavable
Color Transparent/White
Disposable Reusable
For Use With All Pipettes with Tips Mounted
Material Silicone
Number of Channels 1
Sterility Non-sterile
Type Single Channel Pipette Adapter
For Use With (Application) Testing with Tips
For Use With (Equipment) PLT Pipette Leak Testing Unit
Includes Plug
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.