missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Accessories for PLT™ (Pipette Leak Testing) Units
Shop All Brand Products
Click to view available options
Type:
Multichannel Pipet Adapter
Replacement Filters
Single Channel Pipette Adapter
Universal Power Supply
For Use With (Application):
Testing with Tips
Testing with or without Tips
Testing without Tips
Unit Size:
Each
Pack of 10
Description
Available accessories include:
- Part No. 703978: Polyethylene filters and cap for pipette adapters, includes ten filters and one cap.
- Part No. 703977: Four-channel pipette adapter, for testing with and without tips, includes four plugs.
- Part No. 703975: Single-channel pipette adapter for testing with tips, includes one plug.
- Part No. 703976: Single-channel pipette adapter for testing without tips, includes one plug.
- Part No. 703979: Universal AC Adapter, 100-240V, 50/60Hz.
Specifications
Specifications
| Compatible Tips | All air displacement pipettes (other than 10 mL Gilson without tips) |
| Description | Single Channel Pipette Adapter, For testing without tips, Includes plugs |
| Autoclavable | Not Autoclavable |
| Color | Black |
| Disposable | Reusable |
| For Use With | PLT Unit |
| Material | Silicone |
| Number of Channels | 1 |
| Sterility | Non-sterile |
| Type | Single Channel Pipette Adapter |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction