missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Accessories for PLT™ (Pipette Leak Testing) Units

Product Code. 11648419 Shop All Brand Products
Change view
Click to view available options
Type:
Multichannel Pipet Adapter
Replacement Filters
Single Channel Pipette Adapter
Universal Power Supply
Unit Size:
Each
Pack of 10
For Use With (Application):
Testing with Tips
Testing with or without Tips
Testing without Tips
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize For Use With (Application)
11648419 Replacement Filters Pack of 10 -
11683239 Multichannel Pipet Adapter Each Testing with or without Tips
11693239 Single Channel Pipette Adapter Each Testing with Tips
11673239 Single Channel Pipette Adapter Each Testing without Tips
11633239 Universal Power Supply Each -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11648419 Supplier BRAND™ Supplier No. 703978
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with BRAND™ PLT™ unit. These accessories are designed for improved process reliability in identifying even the smallest leaks in your air displacement pipette.

Available accessories include:

  • Part No. 703978: Polyethylene filters and cap for pipette adapters, includes ten filters and one cap.
  • Part No. 703977: Four-channel pipette adapter, for testing with and without tips, includes four plugs.
  • Part No. 703975: Single-channel pipette adapter for testing with tips, includes one plug.
  • Part No. 703976: Single-channel pipette adapter for testing without tips, includes one plug.
  • Part No. 703979: Universal AC Adapter, 100-240V, 50/60Hz.
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips All tips
Description Polyethylene Replacement Filters
Autoclavable Autoclavable
Color White
Disposable Single-use
For Use With PLT Unit
Material Polyethylene
Number of Channels Variable
Sterility Non-sterile
Type Replacement Filters
For Use With (Equipment) PLT Pipette Leak Testing Unit
Includes 10 filters and 1 stopper
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.