missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ 8-Strip PCR Caps, BIO-CERT™ PCR QUALITY

Product Code. 10385962 Shop All Brand Products
Change view
Click to view available options
Color:
Blue
Clear
Green
Rose
Yellow
For Use With (Equipment):
Microplates
Thermal cycler
qPCR plates
Unit Size:
Pack of 125
Pack of 300
Format:
Domed
Flat
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color For Use With (Equipment) unitSize Format
10385962 Blue Microplates Pack of 125 -
10608131 Clear Thermal cycler Pack of 125 -
10633493 Yellow Microplates Pack of 125 -
10122662 Rose Microplates Pack of 125 -
10539482 Green Microplates Pack of 125 -
10271783 Clear Microplates Pack of 125 Flat
15644007 Clear qPCR plates Pack of 300 Flat
15664007 Clear qPCR plates Pack of 300 Domed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10385962 Supplier BRAND™ Supplier No. 781344
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

BRAND™ PCR cap strips ensure reliable closures for PCR strips and plates, safeguarding your samples against evaporation and contamination.

  • Exceptional Quality and Purity: BIO-CERT™ PCR QUALITY ensures caps are free from human DNA, RNase, DNase, pyrogens, and PCR inhibitors.
  • Secure Sealing: Designed to reliably seal PCR strips and plates with 24-, 48-, or 96-wells, effectively protecting your samples from evaporation and contamination.
  • Perfect Compatibility: Optimally shaped to match BRAND™ PCR tube strips and plates, ensuring easy opening and closing. They are also compatible with most other manufacturers, offering flexibility for your workflow.
  • Contamination Prevention: Equipped with side tabs for precise positioning and a small opening attachment on each cap to prevent direct contact with the interior.
  • qPCR-Ready Transparency: Highly transparent flat caps enable accurate fluorescence measurement, making them ideal for qPCR applications.
  • Enhanced Sealing with Domed Caps: Domed caps provide a stronger sealing effect when used with thermal cyclers lacking pressure sensors.
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
For Use With (Equipment) Microplates
Shape Domed
Sterility Non-sterile
Material Polypropylene
Dnase Free Yes
RNase Free Yes
Closure Type 8-strip Domed Cap
Autoclavable Autoclavable
Closure Included Yes
Disposable Single-use
Product Type Cap Strip
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.