Description
- Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.
- For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
- Available in a variety of thread sizes.
Specifications
Specifications
| Product Type | Dispenser Thread Adapter |
| Material | ETFE |
| For Use With (Equipment) | Dispensette, seripettor, Titrette, QuikSip |
| Quantity | 1 Ea. |
| Thread Size | GL 45/38 |