missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ pureGrade™ S 96-well Microplates

Product Code. 10479183 Shop All Brand Products
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
White, Clear Bottom
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Rounded Square
Unit Size:
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Well Shape unitSize
10479183 Black Flat Bottom (F-well) Pack of 50
10154183 Clear Round Bottom (U-well) Pack of 50
10616953 Clear Conical Bottom (V-well) Pack of 50
10469183 Clear Flat Bottom (F-well) Pack of 50
10091963 Clear Rounded Square Pack of 50
10513054 White Flat Bottom (F-well) Pack of 50
10467863 White, Clear Bottom Flat Bottom (F-well) Pack of 50
10657333 Black, Clear Bottom Flat Bottom (F-well) Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10479183 Supplier BRAND™ Supplier No. 781668
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Nontreated sterile microplates for diverse applications

  • PS surface
  • Medium binding
  • Sterilized via beta-radiation
  • Standard plates available in U-, V-, F-, C-shapes
  • Transparent bottom available in F- shape
  • With lid
  • Available in clear, white or black
  • Gray alphanumeric embossing on white and black solid plates for easy well identification
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic

Bacteriological assays, screenings, storage.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance ISO 11137, AAMI, DIN EN ISO 10993, ANSI, SLAS
Format Standard
Lid With Lid
Array 8 x 12
Binding Type Medium Binding
Color Black
Material Polystyrene
Volume (Metric) Well 350 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) Bacteriological assays, screening and for storage
Quantity 50/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.