missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ pureGrade™ S 384-well Microplates

Product Code. 10153663 Shop All Brand Products
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
White, Clear Bottom
Volume (Metric) Well:
0.1 mL
0.12 mL
Unit Size:
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) Well unitSize
10153663 White 0.1 mL Pack of 50
10721544 Clear 0.1 mL Pack of 50
10617903 Black 0.1 mL Pack of 50
10427493 White, Clear Bottom 0.12 mL Pack of 50
10774034 Black, Clear Bottom 0.12 mL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10153663 Supplier BRAND™ Supplier No. 781681
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Non-treated sterile microplates for diverse applications

  • Non-treated PS surface
  • Medium binding
  • Sterilized via beta-radiation
  • Sterile plate for most applications
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Lid With Lid
Array 16 x 24
Color White
Material Polystyrene
Volume (Metric) Well 0.1 mL
No. of Wells 384
Sterility Sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) Laboratory tasks, such as homogeneous assays, screening and storage
Quantity 50/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.