missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates™ pureGrade™ S 384-well Microplates
Non-treated sterile microplates for diverse applications
Brand: BRAND™ 781687
749.25 GBP valid until 2024-12-31
Use promo code "21894" to get your promotional price.
Code : 03
Additional Details : Weight : 3.75000kg
Description
- Non-treated PS surface
- Medium binding
- Sterilized via beta-radiation
- Sterile plate for most applications
Specifications
Standard | |
16 x 24 | |
Black, Clear Bottom | |
0.12 mL | |
Sterile | |
Flat Bottom (F-well) | |
50/Cs. |
With Lid | |
Individually Wrapped | |
Polystyrene | |
384 | |
Non-treated | |
Laboratory tasks, such as homogeneous assays, screening and storage |