missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ pureGrade™ S 1536-well Microplates

Product Code. 10183563 Shop All Brand Products
Change view
Click to view available options
Color:
Black
Clear
White
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10183563 White Pack of 50
10688463 Clear Pack of 50
10513244 Black Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10183563 Supplier BRAND™ Supplier No. 781701

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Non-treated sterile microplates for diverse applications

  • Nontreated PS surface
  • Medium binding
  • Sterilized via beta-radiation
  • Sterile plate for most applications
  • With lid
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic

Bacteriological assays, screenings, storage.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 11137, AAMI
Format Standard
Lid With Lid
Array 32 x 48
Packaging Individually Wrapped
Color White
Product Type 1536-Well Cell Culture Multiwell Plates
Material Polystyrene
Volume (Metric) Well 10 μL
No. of Wells 1536
Sterility Sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) Sterile applications, bacteriological assays
Quantity 50/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.