missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ pureGrade™ 96-Well Microplates

Product Code. 10337723 Shop All Brand Products
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
Clear, Clear Bottom
White
White, Clear Bottom
Well Shape:
Conical Bottom (V-well)
Curved Edges, Flat Bottom (C-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Volume (Metric) Well:
330 μL
350 μL
360 μL
Surface Treatment:
Non-treated
pureGrade
Unit Size:
Pack of 100
Pack of 50
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Well Shape Volume (Metric) Well Surface Treatment unitSize
10337723 White, Clear Bottom Flat Bottom (F-well) 330 μL Non-treated Pack of 100
10246473 Clear Round Bottom (U-well) 330 μL Non-treated Pack of 100
10052063 Clear Conical Bottom (V-well) 360 μL Non-treated Pack of 100
10143663 Clear Flat Bottom (F-well) 350 μL Non-treated Pack of 100
10062063 Clear Curved Edges, Flat Bottom (C-well) 350 μL Non-treated Pack of 100
10002213 White Round Bottom (U-well) 330 μL Non-treated Pack of 100
10732694 White Flat Bottom (F-well) 350 μL Non-treated Pack of 100
10448433 Black Round Bottom (U-well) 330 μL Non-treated Pack of 100
10742694 Black Flat Bottom (F-well) 350 μL Non-treated Pack of 100
10347723 Black, Clear Bottom Flat Bottom (F-well) 330 μL Non-treated Pack of 100
10004804 Clear, Clear Bottom Flat Bottom (F-well) 350 μL Non-treated Pack of 50
15674007 Black Flat Bottom (F-well) 350 μL pureGrade Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 10337723 Supplier BRAND™ Supplier No. 781610
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Constructed of polystyrene with nontreated surface. BRANDplates™ pureGrade™ 96-Well Microplates are noncytotoxic, and free from endotoxins, DNase, DNA, and RNase. Medium binding, nonsterile microplates are suited for homogenous assays, screening, and storage with gray alphanumeric embossing on white and black solid plates for easy well identification.

  • Polystyrene surface
  • Medium binding, nonsterile
  • Standard plate for most applications
  • Gray alphanumeric embossing on white and black solid plates for easy well identification
  • Strip plates available
  • Standard plates available in U-, V-, F-, C-shapes
  • Transparent bottoms available in F- shape
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic
  • Suited for homogenous assays, screening, and storage.
TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance ANSI, SLAS
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Medium Binding
Color White, Clear Bottom
Product Type Transparent bottom
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Homogenous assays, screenings and storage
Quantity 100/Cs.
RNase Free Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.