missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates™ pureGrade™ 1536-Well Microplates
Shop All Brand Products
Click to view available options
Color:
Black
Clear
White
Unit Size:
Pack of 50
Description
- Nontreated PS surface
- Medium binding, nonsterile
- Standard plate for most applications
- Available in clear, white or black
- Free from endotoxins, DNase, DNA, RNase; noncytotoxic
Homogenous assays, screening, and storage.
Specifications
Specifications
| Barcode | No Barcode |
| Binding Property | Medium Binding |
| Certifications/Compliance | ISO 11137, AAMI |
| Format | Standard |
| Lid | Without Lid |
| Array | 32 x 48 |
| Autoclavable | Not Autoclavable |
| Binding Type | Untreated |
| Color | White |
| Disposable | Single-use |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction