missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- High binding for molecules with mixed hydrophilic and hydrophobic regions
- Surface choice for majority of standard ELISAs
- For solid phase immunoassays
- PS, non-sterile
- Available in clear, white or black
- Free from endotoxins, DNase, DNA, RNase, non-cytotoxic
Specifications
Specifications
| Barcode | No Barcode |
| Format | Standard |
| Lid | With Lid |
| Array | 16 x 24 |
| Color | Black |
| Material | Polystyrene |
| Volume (Metric) Well | 100 μL |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| Surface Treatment | immunoGrade |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?