missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ immunoGrade™ 384-Well Microplates

Product Code. p-4830986 Shop All Brand Products
Change view
Click to view available options
Color:
Black
Clear
White
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10082253 Clear Pack of 50
10296803 White Pack of 50
10164093 Black Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10082253 Supplier BRAND™ Supplier No. 781740
Add to basket
Add to basket
This item is not returnable. View return policy

Immunoassay plate optimized for immobilization of IgG

  • High binding for molecules with mixed hydrophilic and hydrophobic regions
  • Surface choice for majority of standard ELISAs
  • For solid phase immunoassays
  • PS, non-sterile
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase, non-cytotoxic
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Lid Without Lid
Array 16 x 24
Color Clear
Material Polystyrene
Volume (Metric) Well 100 μL
No. of Wells 384
Sterility Non-sterile
Surface Treatment immunoGrade
Well Shape Flat Bottom (F-well)
For Use With (Application) For immobilization of IgG, solid phase immunoassays
Quantity 50/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.