missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ White Real-Time PCR (qPCR) 96-Well Microplates, BIO-CERT™ PCR QUALITY, Standard Profile

Product Code. p-4122342 Shop All Brand Products
Change view
Click to view available options
Includes:
Half Skirt
No Skirt
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
10034564 No Skirt Pack of 50
10696095 Half Skirt Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10034564 Supplier BRAND™ Supplier No. 781354
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed optimized results when performing quantitative Real Time PCR (qPCR)

Designed for optimal performance in fluorescence signal analysis and DNA quantification, these plates are ideal for qPCR applications. Key features include:

  • Enhanced Signal Reflection: White surface and titanium dioxide (TiO2) coloring maximize fluorescence signal reflection for precise quantification.
  • Exceptional Purity: BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors.
  • Advanced Manufacturing Standards: Produced in state-of-the-art clean room conditions for consistent quality.
  • Superior Thermal Performance: Extra-thin walls provide efficient thermal transfer.
  • User-Friendly Design:
    • Smooth surfaces optimize signal reflection
    • Blue embossed alphanumeric coding and a cut corner design ensure easy orientation
    • Standard-profile, with an elevated rim offers compatibility with various thermal cyclers
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Untreated
Certifications/Compliance DNA, DNase, RNase-free
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Autoclavable
Packaging Bagged
Color White
Disposable Single-use
Product Type qPCR 96-well plate
For Use With (Equipment) Thermal Cyclers
Skirt Style Non-skirted
Material Polypropylene
Volume (Metric) Well 0.20 mL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 0.2 mL
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) Optimize results when performing quantitative RT-PCR
Includes No Skirt
No. per Pack 50/Pk.
Profile Standard
RNase Free Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.