Learn More
BRAND™ Replacement Dispenser Cartridges
Replacement Cartridges for seripettor™ and seripettor™ pro, and QuickSip bottletop dispensers
Brand: BRAND™ 704500
Additional Details : Weight : 0.01000kg
Description
- Sterile or non-sterile
- For use with seripettor™, seripettor™ pro and QuickSip bottletop dispensers
Specifications
Dispensing Cartridge | |
2 mL | |
Non-sterile |
Polyethylene, Polypropylene | |
Seripettor™ / Seripettor™ pro / QuikSip |